Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163548.1 | NP_001157020.1 | 3 - 4 | 705 | 72 | |
NM_011182.4 | NP_035312.3 | 3 - 4 | 381 | 72 | ||
GenBank mRNA | AB013470.1 | - | 3 - 4 | 235 | 72 | |
AF084221.1 | - | 3 - 4 | 275 | 72 | ||
AK016645.1 | - | 3 - 4 | 707 | 72 | ||
AK036191.1 | - | 3 - 4 | 295 | 72 | ||
AK142407.1 | - | 3 - 4 | 370 | 72 | ||
AK159229.1 | - | 3 - 4 | 353 | 72 | ||
AK168566.1 | - | 3 - 4 | 295 | 72 | ||
AK220565.1 | - | 3 - 4 | 361 | 72 | ||
BC007189.1 | - | 2 - 3 | 122 | 72 | ||
BC035296.1 | - | 3 - 4 | 322 | 3969080 | 72 | |
BY271755.1 | - | 3 - 4 | 381 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |