Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083935.1 | NP_001077404.1 | 7 - 8 | 918 | 85 | |
NM_001083936.1 | NP_001077405.1 | 6 - 7 | 951 | 85 | ||
NM_011199.2 | NP_035329.2 | 8 - 9 | 920 | 85 | ||
GenBank mRNA | BC013446.1 | - | 8 - 9 | 897 | 4241234 | 85 |
BC051981.1 | - | 6 - 7 | 822 | 6402767 | 85 | |
X78936.1 | - | 6 - 7 | 697 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |