Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039363.2 | NP_001034452.1 | 35 - 36 | 4776 | 58 | |
NM_001286713.1 | NP_001273642.1 | 37 - 38 | 4974 | 58 | ||
NM_001286714.1 | NP_001273643.1 | 36 - 37 | 4900 | 58 | ||
NM_001286716.1 | NP_001273645.1 | 35 - 36 | 4771 | 58 | ||
NM_001286718.1 | NP_001273647.1 | 35 - 36 | 4773 | 58 | ||
NM_001286720.1 | NP_001273649.1 | 37 - 38 | 4974 | 58 | ||
NM_011647.3 | NP_035777.2 | 36 - 37 | 4905 | 58 | ||
GenBank mRNA | AF132986.1 | - | 33 - 34 | 4499 | 58 | |
AK143799.1 | - | 35 - 36 | 4774 | 58 | ||
AK147302.1 | - | 36 - 37 | 4846 | 58 | ||
AK147487.1 | - | 37 - 38 | 4943 | 58 | ||
AK147874.1 | - | 35 - 36 | 4740 | 58 | ||
AK147908.1 | - | 36 - 37 | 4906 | 58 | ||
AK148062.1 | - | 36 - 37 | 4897 | 58 | ||
AK148271.1 | - | 23 - 24 | 3345 | 58 | ||
AK163991.1 | - | 1 - 2 | 124 | 58 | ||
AK164586.1 | - | 1 - 2 | 54 | 58 | ||
AK170589.1 | - | 35 - 36 | 4767 | 58 | ||
BC052449.1 | - | 35 - 36 | 4752 | 6409037 | 58 | |
BC060701.1 | - | 35 - 36 | 4752 | 6409037 | 58 | |
U37775.1 | - | 35 - 36 | 4817 | 58 | ||
U39818.1 | - | 37 - 38 | 4942 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |