Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011976.1 | NP_036106.1 | 6 - 7 | 1029 | 76 | |
GenBank mRNA | AF134918.1 | - | 6 - 7 | 1029 | 76 | |
BC049183.1 | - | 6 - 7 | 1022 | 6395010 | 76 | |
BC054532.1 | - | 7 - 8 | 1067 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |