Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139509.1 | NP_001132981.1 | 2 - 3 | 303 | 77 | |
NM_013613.2 | NP_038641.1 | 2 - 3 | 449 | 77 | ||
GenBank mRNA | AK087495.1 | - | 3 - 4 | 324 | 77 | |
BC137715.1 | - | 2 - 3 | 761 | 8860734 | 77 | |
BC144854.1 | - | 2 - 3 | 977 | 77 | ||
S53744.1 | - | 2 - 3 | 340 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |