Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013708.4 | NP_038736.1 | 2 - 3 | 424 | 79 | |
GenBank mRNA | AF148129.1 | - | 2 - 3 | 286 | 79 | |
AF204053.1 | - | 2 - 3 | 424 | 79 | ||
AK044718.1 | - | 2 - 3 | 456 | 79 | ||
AK053560.1 | - | 2 - 3 | 392 | 79 | ||
BC017521.1 | - | 2 - 3 | 418 | 4512088 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |