Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164155.1 | NP_001157627.1 | 7 - 8 | 895 | 66 | |
NM_013869.5 | NP_038897.4 | 7 - 8 | 1498 | 66 | ||
GenBank mRNA | AB040432.1 | - | 7 - 8 | 859 | 66 | |
AF167552.1 | - | 7 - 8 | 804 | 66 | ||
AF173166.1 | - | 6 - 7 | 760 | 66 | ||
AK028146.1 | - | 7 - 8 | 805 | 66 | ||
AK050262.1 | - | 3 - 4 | 464 | 66 | ||
AK083283.1 | - | 7 - 8 | 1113 | 66 | ||
AK181271.1 | - | 1 - 2 | 55 | 66 | ||
AK192918.1 | - | 1 - 2 | 55 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mus musculus Probe spans exons |