Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013920.2 | NP_038948.1 | 2 - 3 | 470 | 80 | |
GenBank mRNA | AJ242626.1 | - | 2 - 3 | 422 | 80 | |
AK143816.1 | - | 2 - 3 | 471 | 80 | ||
BC125519.1 | - | 2 - 3 | 292 | 40130034 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |