Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163336.1 | NP_001156808.1 | 11 - 12 | 1601 | 113 | |
NM_001163337.1 | NP_001156809.1 | 11 - 12 | 1601 | 113 | ||
NM_016745.3 | NP_058025.2 | 11 - 12 | 1601 | 113 | ||
GenBank mRNA | AK089487.1 | - | 11 - 12 | 1601 | 113 | |
AK155904.1 | - | 11 - 12 | 1602 | 113 | ||
BC017639.1 | - | 11 - 12 | 1515 | 4924519 | 113 | |
BC026147.1 | - | 11 - 12 | 1547 | 4191458 | 113 | |
U49393.1 | - | 11 - 12 | 1462 | 113 | ||
U49394.1 | - | 11 - 12 | 1462 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |