Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019488.4 | NP_062361.1 | 7 - 8 | 1020 | 85 | |
GenBank mRNA | AF232061.2 | - | 7 - 8 | 977 | 85 | |
AJ245936.1 | - | 7 - 8 | 997 | 85 | ||
AK081806.1 | - | 7 - 8 | 1037 | 85 | ||
AK155792.1 | - | 7 - 8 | 1049 | 85 | ||
AK170319.1 | - | 7 - 8 | 1033 | 85 | ||
AY856043.1 | - | 7 - 8 | 981 | 85 | ||
BC021758.1 | - | 3 - 4 | 346 | 85 | ||
BC090993.1 | - | 7 - 8 | 1020 | 6751763 | 85 | |
Y17802.1 | - | 7 - 8 | 1002 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |