Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159564.1 | NP_001153036.1 | 5 - 6 | 482 | 79 | |
NM_021359.3 | NP_067334.1 | 2 - 3 | 376 | 79 | ||
GenBank mRNA | AF115376.1 | - | 5 - 6 | 328 | 79 | |
AK008867.1 | - | 2 - 3 | 377 | 79 | ||
AK036439.1 | - | 5 - 6 | 483 | 79 | ||
AK085900.1 | - | 5 - 6 | 523 | 79 | ||
BC049185.1 | - | 2 - 3 | 345 | 6475423 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |