Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jarid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205043.1 | NP_001191972.1 | 12 - 13 | 3447 | 74 | |
NM_001205044.1 | NP_001191973.1 | 13 - 14 | 3120 | 74 | ||
NM_021878.3 | NP_068678.1 | 13 - 14 | 3166 | 74 | ||
GenBank mRNA | AK045214.1 | - | 12 - 13 | 3252 | 74 | |
AK147226.1 | - | 12 - 13 | 3447 | 74 | ||
AK164134.1 | - | 11 - 12 | 2771 | 74 | ||
BC003374.1 | - | 6 - 7 | 1016 | 74 | ||
BC026056.1 | - | 12 - 13 | 3354 | 74 | ||
BC052444.1 | - | 13 - 14 | 3037 | 6406875 | 74 | |
BC060695.1 | - | 13 - 14 | 3037 | 6406875 | 74 | |
BC115758.1 | - | 7 - 8 | 2051 | 74 | ||
D31967.1 | - | 13 - 14 | 3056 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |