Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 5 - 6 | 1052 | 106 | |
GenBank mRNA | AF131197.1 | - | 5 - 6 | 994 | 106 | |
AK028478.1 | - | 5 - 6 | 1053 | 106 | ||
BC071215.1 | - | 5 - 6 | 1016 | 4196138 | 106 | |
U18084.1 | - | 1 - 2 | 155 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |