Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199188.1 | NP_001186117.1 | 9 - 10 | 1026 | 91 | |
NM_024225.5 | NP_077187.1 | 8 - 9 | 937 | 91 | ||
GenBank mRNA | AK003091.1 | - | 650 | 91 | ||
AK007676.1 | - | 8 - 9 | 885 | 91 | ||
AK029463.1 | - | 8 - 9 | 897 | 91 | ||
AK031873.1 | - | 9 - 10 | 918 | 91 | ||
AK049129.1 | - | 9 - 10 | 918 | 91 | ||
AK078776.1 | - | 9 - 10 | 919 | 91 | ||
AK144710.1 | - | 9 - 10 | 922 | 91 | ||
AK152311.1 | - | 9 - 10 | 963 | 91 | ||
AK159115.1 | - | 5 - 6 | 480 | 91 | ||
AK159904.1 | - | 8 - 9 | 842 | 91 | ||
AK167362.1 | - | 8 - 9 | 857 | 91 | ||
AK169545.1 | - | 7 - 8 | 1832 | 91 | ||
AK169786.1 | - | 9 - 10 | 918 | 91 | ||
AK212372.1 | - | 3 - 4 | 252 | 91 | ||
AK214918.1 | - | 3 - 4 | 277 | 91 | ||
BC002242.1 | - | 9 - 10 | 995 | 3492262 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |