Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172481.1 | NP_001165952.1 | 4 - 5 | 861 | 72 | |
NM_025711.3 | NP_079987.2 | 4 - 5 | 878 | 72 | ||
GenBank mRNA | AF316825.1 | - | 3 - 4 | 598 | 72 | |
AK014504.1 | - | 4 - 5 | 834 | 72 | ||
AK085920.1 | - | 4 - 5 | 833 | 72 | ||
BC034888.1 | - | 4 - 5 | 780 | 1365428 | 72 | |
BC145904.1 | - | 4 - 5 | 776 | 40131207 | 72 | |
BC145930.1 | - | 4 - 5 | 759 | 40131233 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |