Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053198.3 | NP_444428.3 | 6 - 7 | 381 | 67 | |
GenBank mRNA | AF325263.1 | - | 6 - 7 | 323 | 67 | |
BC089535.1 | - | 6 - 7 | 363 | 6589805 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |