Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133661.3 | NP_598422.2 | 13 - 14 | 1844 | 122 | |
GenBank mRNA | BC019211.1 | - | 13 - 14 | 1848 | 5053116 | 122 |
M97632.1 | - | 13 - 14 | 1584 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |