Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010348.3 | NP_034478.1 | 11 - 12 | 2306 | 89 | |
NM_146072.4 | NP_666184.2 | 11 - 12 | 2306 | 89 | ||
GenBank mRNA | AK042635.1 | - | 11 - 12 | 2307 | 89 | |
AK043542.1 | - | 11 - 12 | 2286 | 89 | ||
AK043737.1 | - | 11 - 12 | 2270 | 89 | ||
AK044250.1 | - | 11 - 12 | 2265 | 89 | ||
AK044420.1 | - | 11 - 12 | 2261 | 89 | ||
AK133667.1 | - | 11 - 12 | 2292 | 89 | ||
AK194777.1 | - | 1 - 2 | 192 | 89 | ||
BC031822.1 | - | 10 - 11 | 1728 | 4501004 | 89 | |
BC049275.1 | - | 10 - 11 | 1728 | 4501004 | 89 | |
X66118.1 | - | 11 - 12 | 2292 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |