Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009388.6 | NP_033414.1 | 10 - 11 | 1539 | 59 | |
GenBank mRNA | AF195533.1 | - | 9 - 10 | 1205 | 59 | |
AK002627.1 | - | 10 - 11 | 1492 | 59 | ||
AK012794.1 | - | 10 - 11 | 1494 | 59 | ||
AK030446.1 | - | 10 - 11 | 1522 | 59 | ||
AK140965.1 | - | 10 - 11 | 1523 | 59 | ||
AK144146.1 | - | 10 - 11 | 1494 | 59 | ||
AK145677.1 | - | 10 - 11 | 1492 | 59 | ||
AK146157.1 | - | 10 - 11 | 1492 | 59 | ||
AK150139.1 | - | 10 - 11 | 1492 | 59 | ||
AK150769.1 | - | 10 - 11 | 1488 | 59 | ||
AK150844.1 | - | 10 - 11 | 1488 | 59 | ||
AK150856.1 | - | 10 - 11 | 1488 | 59 | ||
AK150901.1 | - | 10 - 11 | 1491 | 59 | ||
AK151916.1 | - | 10 - 11 | 1487 | 59 | ||
AK151969.1 | - | 10 - 11 | 1487 | 59 | ||
AK152245.1 | - | 10 - 11 | 1487 | 59 | ||
AK152460.1 | - | 10 - 11 | 1508 | 59 | ||
AK152886.1 | - | 10 - 11 | 1492 | 59 | ||
AK159922.1 | - | 10 - 11 | 1492 | 59 | ||
AK166763.1 | - | 10 - 11 | 1526 | 59 | ||
AK167084.1 | - | 10 - 11 | 1526 | 59 | ||
AK205198.1 | - | 2 - 3 | 189 | 59 | ||
BC055336.1 | - | 10 - 11 | 1466 | 6413263 | 59 | |
U05809.1 | - | 10 - 11 | 1487 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |