Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 14 - 15 | 1592 | 108 | |
GenBank mRNA | AF057367.1 | - | 14 - 15 | 1572 | 108 | |
AK142512.1 | - | 14 - 15 | 1508 | 108 | ||
AK155922.1 | - | 14 - 15 | 1337 | 108 | ||
AK162410.1 | - | 12 - 13 | 1165 | 108 | ||
BC053061.1 | - | 14 - 15 | 1575 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |