Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 20 - 21 | 2909 | 116 | |
GenBank mRNA | AF057367.1 | - | 20 - 21 | 2889 | 116 | |
AK142512.1 | - | 20 - 21 | 2825 | 116 | ||
AK162410.1 | - | 18 - 19 | 2482 | 116 | ||
AK164917.1 | - | 7 - 8 | 1354 | 116 | ||
AK220463.1 | - | 21 - 22 | 2866 | 116 | ||
BC053061.1 | - | 20 - 21 | 2891 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |