Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011172.2 | NP_035302.2 | 6 - 7 | 883 | 87 | |
GenBank mRNA | AF120279.1 | - | 6 - 7 | 840 | 87 | |
AK144068.1 | - | 6 - 7 | 853 | 87 | ||
BC008548.1 | - | 6 - 7 | 707 | 87 | ||
BC125327.1 | - | 6 - 7 | 686 | 40129842 | 87 | |
U80020.1 | - | 6 - 7 | 832 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |