Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013875.5 | NP_038903.3 | 3 - 4 | 545 | 118 | |
GenBank mRNA | AF190639.1 | - | 3 - 4 | 326 | 118 | |
AJ251859.1 | - | 3 - 4 | 597 | 118 | ||
AK035385.1 | - | 3 - 4 | 557 | 118 | ||
BC130267.1 | - | 3 - 4 | 426 | 40143060 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |