Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700023087.mm.1 |
TC0700002659.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023087.mm.1 |
NM_001046637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023087.mm.1 |
NM_001046637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023087.mm.1 |
NM_001285825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023087.mm.1 |
NM_015782.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023087.mm.1 |
NM_015782.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023084.mm.1 |
TC0700002659.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023084.mm.1 |
NM_001046637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023084.mm.1 |
NM_001046637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023084.mm.1 |
NM_001285825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023084.mm.1 |
NM_015782.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023084.mm.1 |
NM_015782.3 |
Refseq |