Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018857.1 | NP_061345.1 | 12 - 13 | 1487 | 107 | |
GenBank mRNA | AK144391.1 | - | 13 - 14 | 1419 | 107 | |
AK146799.1 | - | 12 - 13 | 1537 | 107 | ||
BC023753.1 | - | 12 - 13 | 1491 | 5354484 | 107 | |
D86370.1 | - | 12 - 13 | 1487 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |