Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033988.2 | NP_001029160.1 | 3 - 4 | 419 | 144 | |
NM_019744.4 | NP_062718.2 | 3 - 4 | 329 | 144 | ||
GenBank mRNA | AF159461.1 | - | 3 - 4 | 326 | 144 | |
AK032951.1 | - | 1 - 2 | 85 | 144 | ||
AK129020.1 | - | 3 - 4 | 377 | 144 | ||
AK132981.1 | - | 144 | ||||
AK143965.1 | - | 3 - 4 | 324 | 144 | ||
AK145070.1 | - | 3 - 4 | 380 | 144 | ||
AK145945.1 | - | 3 - 4 | 372 | 144 | ||
AK145954.1 | - | 3 - 4 | 376 | 144 | ||
AK146384.1 | - | 3 - 4 | 330 | 144 | ||
AK146498.1 | - | 3 - 4 | 369 | 144 | ||
AK146598.1 | - | 3 - 4 | 353 | 144 | ||
AK146646.1 | - | 3 - 4 | 324 | 144 | ||
AK146649.1 | - | 3 - 4 | 326 | 144 | ||
AK147036.1 | - | 3 - 4 | 327 | 144 | ||
AK147125.1 | - | 3 - 4 | 371 | 144 | ||
BC031528.1 | - | 3 - 4 | 370 | 4008986 | 144 | |
BC085086.1 | - | 3 - 4 | 315 | 6335192 | 144 | |
CF161810.1 | - | 3 - 4 | 414 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |