Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019938.3 | NP_064322.1 | 14 - 15 | 2226 | 73 | |
GenBank mRNA | AB029919.1 | - | 14 - 15 | 2226 | 73 | |
AK006021.1 | - | 5 - 6 | 677 | 73 | ||
AK133053.1 | - | 13 - 14 | 2281 | 73 | ||
BC138245.1 | - | 14 - 15 | 2178 | 8861264 | 73 | |
BC138246.1 | - | 14 - 15 | 2178 | 8861265 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |