Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025571.2 | NP_001020742.1 | 3 - 4 | 690 | 81 | |
NM_021309.3 | NP_067284.1 | 3 - 4 | 690 | 81 | ||
GenBank mRNA | AF203343.1 | - | 3 - 4 | 697 | 81 | |
AK037489.1 | - | 3 - 4 | 412 | 81 | ||
AK172182.1 | - | 3 - 4 | 442 | 81 | ||
BC034847.1 | - | 3 - 4 | 422 | 3327113 | 81 | |
U69490.1 | - | 3 - 4 | 400 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |