Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
NM_001127351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
NM_001127351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
NM_001177804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
NM_001177804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
NM_022433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
NM_022433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037873.mm.1 |
TC0700004509.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
NM_001127351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
NM_001127351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
NM_001177804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
NM_001177804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
NM_022433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
NM_022433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037874.mm.1 |
TC0700004509.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037881.mm.1 |
NM_022433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037881.mm.1 |
NM_022433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037881.mm.1 |
TC0700004509.mm.1 |
MTA TC |