Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacsin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289677.1 | NP_001276606.1 | 2 - 3 | 268 | 96 | |
NM_001289678.1 | NP_001276607.1 | 3 - 4 | 267 | 96 | ||
NM_028733.4 | NP_083009.1 | 3 - 4 | 397 | 96 | ||
NM_030880.3 | NP_112019.2 | 3 - 4 | 263 | 96 | ||
GenBank mRNA | AF149824.1 | - | 3 - 4 | 181 | 96 | |
AF242531.1 | - | 2 - 3 | 166 | 96 | ||
AK049166.1 | - | 3 - 4 | 398 | 96 | ||
AK141063.1 | - | 3 - 4 | 240 | 96 | ||
BC003884.1 | - | 3 - 4 | 204 | 3497435 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |