Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031868.2 | NP_114074.1 | 5 - 6 | 787 | 100 | |
GenBank mRNA | AK007932.1 | - | 5 - 6 | 788 | 100 | |
AK028392.1 | - | 5 - 6 | 777 | 100 | ||
AK090070.1 | - | 5 - 6 | 805 | 100 | ||
AK150344.1 | - | 5 - 6 | 805 | 100 | ||
AK151582.1 | - | 5 - 6 | 805 | 100 | ||
AK152051.1 | - | 5 - 6 | 805 | 100 | ||
AK152585.1 | - | 5 - 6 | 805 | 100 | ||
AK152667.1 | - | 5 - 6 | 783 | 100 | ||
AK153517.1 | - | 5 - 6 | 769 | 100 | ||
AK158681.1 | - | 5 - 6 | 743 | 100 | ||
AK159575.1 | - | 5 - 6 | 795 | 100 | ||
AK167244.1 | - | 5 - 6 | 788 | 100 | ||
AK167538.1 | - | 5 - 6 | 800 | 100 | ||
AK167880.1 | - | 5 - 6 | 789 | 100 | ||
AK167981.1 | - | 5 - 6 | 788 | 100 | ||
BC014828.1 | - | 5 - 6 | 793 | 4239005 | 100 | |
U25809.1 | - | 5 - 6 | 808 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |