Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146239.2 | NP_666351.2 | 13 - 14 | 1680 | 110 | |
GenBank mRNA | BC031778.1 | - | 1227 | 4501115 | 110 | |
BC049904.1 | - | 1227 | 4501115 | 110 | ||
BC064815.1 | - | 13 - 14 | 1665 | 6516981 | 110 | |
BQ442862.1 | - | 3 - 4 | 264 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |