Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008230.6 | NP_032256.3 | 7 - 8 | 920 | 78 | |
GenBank mRNA | AF109137.1 | - | 7 - 8 | 830 | 78 | |
AK076384.1 | - | 6 - 7 | 2892 | 78 | ||
AK088545.1 | - | 7 - 8 | 907 | 78 | ||
AK133455.1 | - | 7 - 8 | 921 | 78 | ||
AK150168.1 | - | 7 - 8 | 908 | 78 | ||
AK153104.1 | - | 7 - 8 | 908 | 78 | ||
BC052833.1 | - | 7 - 8 | 905 | 30044098 | 78 | |
X57437.1 | - | 7 - 8 | 885 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |