Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019429.2 | NP_062302.1 | 3 - 4 | 376 | 155 | |
GenBank mRNA | AJ131775.1 | - | 3 - 4 | 376 | 155 | |
AK037514.1 | - | 2 - 3 | 451 | 155 | ||
AK088019.1 | - | 3 - 4 | 404 | 155 | ||
BC125481.1 | - | 3 - 4 | 374 | 40129996 | 155 | |
BC131955.1 | - | 3 - 4 | 374 | 40130232 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |