Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081695.2 | NP_001075164.1 | 9 - 10 | 988 | 76 | |
NM_001284197.1 | NP_001271126.1 | 7 - 8 | 829 | 76 | ||
NM_019448.4 | NP_062321.1 | 8 - 9 | 842 | 76 | ||
GenBank mRNA | AF220524.1 | - | 7 - 8 | 773 | 76 | |
AJ404467.1 | - | 7 - 8 | 814 | 76 | ||
AK010434.1 | - | 8 - 9 | 843 | 76 | ||
BC083147.1 | - | 7 - 8 | 827 | 30447906 | 76 | |
EF051624.1 | - | 9 - 10 | 988 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |