Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053253.3 | NP_444483.2 | 8 - 9 | 1022 | 77 | |
GenBank mRNA | AF123386.1 | - | 6 - 7 | 653 | 77 | |
AK132332.1 | - | 8 - 9 | 1031 | 77 | ||
BC052357.1 | - | 8 - 9 | 1022 | 6772144 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |