Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080467.3 | NP_536715.3 | 7 - 8 | 916 | 115 | |
GenBank mRNA | AB050903.1 | - | 6 - 7 | 642 | 115 | |
AF326316.1 | - | 7 - 8 | 710 | 115 | ||
AF435090.1 | - | 7 - 8 | 904 | 115 | ||
AK128892.1 | - | 7 - 8 | 916 | 115 | ||
BC046979.1 | - | 8 - 9 | 938 | 6315485 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |