Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130447.3 | NP_569714.2 | 6 - 7 | 1527 | 127 | |
GenBank mRNA | AB052157.1 | - | 6 - 7 | 1246 | 127 | |
AF345951.1 | - | 6 - 7 | 1371 | 127 | ||
AF345952.1 | - | 5 - 6 | 1206 | 127 | ||
BC057321.1 | - | 6 - 7 | 1378 | 6830600 | 127 | |
BC059232.1 | - | 6 - 7 | 1314 | 6409944 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |