Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133217.3 | NP_573480.1 | 9 - 10 | 1341 | 68 | |
GenBank mRNA | AJ290392.1 | - | 8 - 9 | 1184 | 68 | |
BC107006.2 | - | 8 - 9 | 1184 | 40046913 | 68 | |
BC107007.2 | - | 8 - 9 | 1184 | 40046914 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |