Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076676.2 | NP_001070144.1 | 11 - 12 | 1485 | 114 | |
NM_001252486.1 | NP_001239415.1 | 11 - 12 | 1485 | 114 | ||
NM_133247.3 | NP_573510.2 | 11 - 12 | 1485 | 114 | ||
GenBank mRNA | AF383174.1 | - | 11 - 12 | 1474 | 114 | |
AK082251.1 | - | 11 - 12 | 1468 | 114 | ||
AK082943.1 | - | 11 - 12 | 1468 | 114 | ||
AK087467.1 | - | 11 - 12 | 1486 | 114 | ||
AK122442.1 | - | 11 - 12 | 1462 | 114 | ||
AK143844.1 | - | 11 - 12 | 1453 | 114 | ||
BC005506.1 | - | 11 - 12 | 1438 | 114 | ||
BC031366.1 | - | 3 - 4 | 601 | 114 | ||
BC036980.1 | - | 4 - 5 | 685 | 114 | ||
BC067399.1 | - | 10 - 11 | 1327 | 114 | ||
BC089315.1 | - | 11 - 12 | 1431 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |