Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134151.4 | NP_598912.4 | 8 - 9 | 1130 | 74 | |
GenBank mRNA | AK028785.1 | - | 8 - 9 | 992 | 74 | |
AK043837.1 | - | 8 - 9 | 1054 | 74 | ||
AK139579.1 | - | 8 - 9 | 975 | 74 | ||
AK145356.1 | - | 8 - 9 | 1023 | 74 | ||
AK151880.1 | - | 8 - 9 | 982 | 74 | ||
AK169708.1 | - | 8 - 9 | 906 | 74 | ||
AK219154.1 | - | 2 - 3 | 154 | 74 | ||
BC013552.1 | - | 8 - 9 | 1061 | 4236514 | 74 | |
BC022143.1 | - | 8 - 9 | 1025 | 5101324 | 74 | |
BC026615.1 | - | 8 - 9 | 1037 | 4976999 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |