Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199349.1 | NP_001186278.1 | 2 - 3 | 391 | 149 | |
NM_138309.3 | NP_612182.1 | 2 - 3 | 391 | 80 | ||
GenBank mRNA | AK031264.1 | - | 2 - 3 | 148 | 149 | |
AK054364.1 | - | 2 - 3 | 191 | 149 | ||
AK080225.1 | - | 2 - 3 | 147 | 149 | ||
AK166275.1 | - | 2 - 3 | 142 | 149 | ||
AY078163.1 | - | 2 - 3 | 187 | 80 | ||
BC031736.1 | - | 2 - 3 | 166 | 4013657 | 80 | |
BY727540.1 | - | 2 - 3 | 392 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |