Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271353.1 | NP_001258282.1 | 5 - 6 | 771 | 74 | |
NM_144855.3 | NP_659104.1 | 5 - 6 | 774 | 74 | ||
NM_178224.3 | NP_835742.1 | 5 - 6 | 774 | 74 | ||
GenBank mRNA | AK077669.1 | - | 5 - 6 | 735 | 74 | |
AK085692.1 | - | 5 - 6 | 736 | 74 | ||
BC013472.1 | - | 5 - 6 | 700 | 4221477 | 74 | |
BC013480.1 | - | 5 - 6 | 710 | 4239757 | 74 | |
BC022144.1 | - | 5 - 6 | 735 | 74 | ||
BC026595.1 | - | 5 - 6 | 707 | 4974777 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |