Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271353.1 | NP_001258282.1 | 13 - 14 | 1595 | 58 | |
NM_144855.3 | NP_659104.1 | 13 - 14 | 1598 | 58 | ||
NM_178224.3 | NP_835742.1 | 13 - 14 | 1598 | 58 | ||
GenBank mRNA | AK077669.1 | - | 13 - 14 | 1559 | 58 | |
BC013472.1 | - | 13 - 14 | 1524 | 4221477 | 58 | |
BC013480.1 | - | 13 - 14 | 1534 | 4239757 | 58 | |
BC022144.1 | - | 13 - 14 | 1559 | 58 | ||
BC026595.1 | - | 13 - 14 | 1531 | 4974777 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |