Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145147.5 | NP_660129.2 | 4 - 5 | 682 | 54 | |
GenBank mRNA | AJ293619.1 | - | 3 - 4 | 312 | 54 | |
AK082192.1 | - | 1 - 2 | 558 | 54 | ||
AK144812.1 | - | 4 - 5 | 683 | 54 | ||
AK152225.1 | - | 4 - 5 | 683 | 54 | ||
AK152972.1 | - | 4 - 5 | 683 | 54 | ||
AK159462.1 | - | 4 - 5 | 655 | 54 | ||
BC027821.1 | - | 1 - 2 | 213 | 5321725 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |