Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031271.1 | - | 10 - 11 | 1156 | 70 | |
AK044016.1 | - | 10 - 11 | 1150 | 70 | ||
AK054393.1 | - | 10 - 11 | 1168 | 70 | ||
AK078023.1 | - | 10 - 11 | 1149 | 70 | ||
AK141216.1 | - | 10 - 11 | 1168 | 70 | ||
AK147680.1 | - | 10 - 11 | 1832 | 70 | ||
AK154520.1 | - | 10 - 11 | 1162 | 70 | ||
BC029082.1 | - | 10 - 11 | 1146 | 3498101 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |