Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153122.2 | NP_694762.1 | 21 - 22 | 3091 | 64 | |
GenBank mRNA | AK164524.1 | - | 5 - 6 | 715 | 64 | |
BC025120.1 | - | 8 - 9 | 1189 | 4020691 | 64 | |
BC034522.1 | - | 21 - 22 | 3091 | 4236444 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |