Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153391.2 | NP_700440.2 | 5 - 6 | 467 | 131 | |
GenBank mRNA | AK043836.1 | - | 5 - 6 | 424 | 131 | |
AK052303.1 | - | 5 - 6 | 447 | 131 | ||
AK147970.1 | - | 5 - 6 | 467 | 131 | ||
AK166172.1 | - | 5 - 6 | 433 | 131 | ||
AY029258.1 | - | 5 - 6 | 537 | 131 | ||
BC060134.1 | - | 5 - 6 | 447 | 131 | ||
BC082301.1 | - | 4 - 5 | 347 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |