Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153391.2 | NP_700440.2 | 13 - 14 | 1412 | 71 | |
GenBank mRNA | AK052303.1 | - | 13 - 14 | 1392 | 71 | |
AK129413.1 | - | 5 - 6 | 501 | 71 | ||
AK147970.1 | - | 13 - 14 | 1412 | 71 | ||
AK166172.1 | - | 13 - 14 | 1378 | 71 | ||
AY029258.1 | - | 13 - 14 | 1482 | 71 | ||
BC060134.1 | - | 13 - 14 | 1406 | 71 | ||
BC082301.1 | - | 12 - 13 | 1292 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |