Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153551.1 | NP_705779.1 | 7 - 8 | 551 | 61 | |
GenBank mRNA | AK014482.1 | - | 7 - 8 | 556 | 61 | |
AK170891.1 | - | 7 - 8 | 549 | 61 | ||
BC035208.1 | - | 7 - 8 | 551 | 3671384 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |